Magnesiumklorid 4,5-hydrat ≥97%, powder, GPR RECTAPUR®

Leverandør: VWR Chemicals

25106.298EA 1150 NOK
25106.298
Magnesiumklorid 4,5-hydrat ≥97%, powder, GPR RECTAPUR®
Magnesiumklorid 4,5-hydrat
Formel: MgCl₂·4,5H₂O
Kokepunkt: 1412 °C (1013 hPa)
Tetthet: 1,57 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
EINECS: 232-094-6

Bestill nå

Spesifikasjon Testresultater

Assay Min. 97 %
Alkaline metals (as SO4) Max. 2.5 %
Heavy metals (as Pb) Max. 20 ppm
SO4 (Sulphate) Max. 0.02 %
Ca (Calcium) Max. 0.1 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR