Saltsyre 36,5 - 38%, GenAR® NF, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

Synonymer: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = R

2515-46EA 0 NOK
2515-46
Saltsyre 36,5 - 38%, GenAR® NF, Macron Fine Chemicals™
Saltsyre
Formel: HCl
MW: 36.46 g/mol
Kokepunkt: 110 °C (1013 hPa)
Smeltepunkt: –30 °C
Tetthet: 1.18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
Store below 25°C
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification Passes Test
NF - Residue on Ignition ≤ 0.008 %
NF - Bromide or Iodide Passes Test
NF - Free Bromine or Chlorine Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Sulfite (SO₃) Passes Test
NF - Assay (HCl)(NF) 36.5 - 38.0 %
Identification A (EP/BP) Passes Test
Identification B (EP/BP) Passes Test
Identification C (EP/BP) Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Free Chlorine (as Cl) ≤ 4 ppm
EP/BP - Sulfate (SO₄) ≤ 20 ppm
EP/BP - Residue after Evaporation ≤ 0.01 %
Assay (HCl)(EP/BP) 35.0 - 39.0 %
JP - Assay (HCl)(JP) 35.0 - 38.0 %
JP - Identification Passes Test
JP - Residue on ignition, mg/10mL ≤ 1.0
JP - Bromide or Iodide Passes Test
JP - Free Bromine or Chlorine Passes Test
JP - Sulfate (SO₄) Passes Test
JP - Sulfite (SO₃) Passes Test
JP - Arsenic (As) ≤ 1 ppm
JP - Heavy Metals (as Pb) (JP) ≤ 5 ppm
JP - Mercury (Hg) ≤ 0.04 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR