Magnesiumsulfat heptahydrat ≥98%, renset

Leverandør: VWR Chemicals

25163.364EA 1175 NOK
25163.364 25163.290 25163.460
Magnesiumsulfat heptahydrat ≥98%, renset
Magnesiumsulfat heptahydrat
Formel: MgSO₄·7H₂O
Smeltepunkt: 1124 °C
Tetthet: 2,66 g/cm³ (20 °C)
MDL nummer: MFCD00149785
CAS-nummer: 10034-99-8
Merck Indeks: 13,05715

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98 %
Insoluble matter Max. 0.05 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR