Mangan (II) klorid tetrahydrat 99.0-101.0%, AnalaR NORMAPUR® ACS analytisk reagens

Leverandør: VWR Chemicals
Warning

25222.233EA 870 NOK
25222.233 25222.290
Mangan (II) klorid tetrahydrat 99.0-101.0%, AnalaR NORMAPUR® ACS analytisk reagens
Mangan (II) klorid tetrahydrat
Formel: MnCl₂·4H₂O
Kokepunkt: 1190 °C (1013 hPa)
Smeltepunkt: 58…59 °C
Tetthet: 2,01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149792
CAS-nummer: 13446-34-9
EINECS: 231-869-6
FN: 3077
ADR: 9,III
Merck Indeks: 13,05751

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 101.0 %
pH (25°C; 5 %) 3.5 - 6.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Not precipitated by (NH4)2S (as SO4) Max. 0.4 %
Reducing substances (as O) Max. 5 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 50 ppm
Co (Cobalt) Max. 20 ppm
Cu (Copper) Max. 5 ppm
K (Potassium) Max. 100 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 50 ppm
Na (Sodium) Max. 0.05 %
Pb (Lead) Max. 5 ppm
Zn (Zinc) Max. 50 ppm
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR