Mangan (II) klorid tetrahydrat 98.0-101.0% (by EDTA titration), krystaller, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Warning

2540-07EA 0 NOK
2540-07 2540-01 2540-R 2540-05
Mangan (II) klorid tetrahydrat 98.0-101.0% (by EDTA titration), krystaller, BAKER ANALYZED® ACS, J.T.Baker®
Mangan (II) klorid tetrahydrat
Formel: MnCl₂·4H₂O
Kokepunkt: 1190 °C (1013 hPa)
Smeltepunkt: 58…59 °C
Tetthet: 2,01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149792
CAS-nummer: 13446-34-9
EINECS: 231-869-6
FN: 3077
ADR: 9,III
Merck Indeks: 13,05751

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements 98.0 - 101.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
ACS - Assay (MnCl₂·4H₂O) (by EDTA titrn) 98.0 - 101.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 3.5 - 6.0
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
ACS - Zinc (Zn) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR