Metylmetakrylat ≥99.5% stabilised, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger


Stabilisering: stabilisert med hydroquinonmonometyleter

25600.296EA 505 NOK
25600.296
Metylmetakrylat ≥99.5% stabilised, GPR RECTAPUR®
Metylmetakrylat
Formel: CH₂=C(CH₃)COOCH₃
Kokepunkt: 100 °C (1013 hPa)
Smeltepunkt: –48 °C
Tetthet: 0,93 g/cm³ (20 °C)
Flash punkt: 8 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008587
CAS-nummer: 80-62-6
EINECS: 201-297-1
FN: 1247
ADR: 3,II
REACH: 01-2119452498-28
Merck Indeks: 12,06005

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.5 %
Acidity Max. 0.0004 meq/g
Colouration Max. 5 APHA
Density (20/4) 0.942 --> 0.944
Stabilizer (MEHQ) 10 --> 25 ppm
Water Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR