Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide), Calbiochem®

Leverandør: Merck Millipore Calbiochem
Warning

256455-1
256455-1EA 9750 NOK
256455-1
Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide), Calbiochem®
Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide)
Formel: C₄₂H₇₅N₃O₁₅
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00161482
CAS-nummer: 86303-23-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR