Nikkel(II)nitrat heksahydrat ≥96%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

25871.292EA 3175 NOK
25871.292
Nikkel(II)nitrat heksahydrat ≥96%, GPR RECTAPUR®
Nikkel(II)nitrat heksahydrat
Formel: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Kokepunkt: 137
Smeltepunkt: 56.7
Tetthet: 2.05 g/cm³ (20 °C)
MDL nummer: MFCD00149805
CAS-nummer: 13478-00-7
EINECS: 236-068-5
FN: 2725
ADR: 5.1 III
Merck Indeks: 12,06596

Bestill nå

Spesifikasjon Testresultater

Assay Min. 96 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.05 %
Cd (Cadmium) Max. 25 ppm
Cu (Copper) Max. 25 ppm
Pb (Lead) Max. 25 ppm
Zn (Zinc) Max. 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR