Sodium tetrachloroaurate dihydrate, TECHNICAL
Leverandør: VWR Chemicals
Warning
Synonymer: Sodium tetrachloroaurate(III) dihydrate
26069.081EA 1800 NOK
26069.081
Sodium tetrachloroaurate dihydrate, TECHNICAL
Sodium tetrachloroaurate dihydrate
Formel: NaAuCl₄·2H₂O
MW: 397.8 g/mol
MDL nummer: MFCD00149162
CAS-nummer: 13874-02-7
EINECS: 239-241-3
FN: 3260
ADR: 8 III
Bestill nå
Spesifikasjon Testresultater
Identification Passes test