Saltsyre 6 mol/l (6 N), AVS TITRINORM Reag. Ph. Eur. 3001500 volumetric solution

Leverandør: VWR Chemicals
Warning

Synonymer: Hydrochloric acid solution, Muriatic acid

2611.5000EA 1075 NOK
2611.5000
Saltsyre 6 mol/l (6 N), AVS TITRINORM Reag. Ph. Eur. 3001500 volumetric solution
Saltsyre
Formel: HCl
Tetthet: 1,06 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 5.976 - 6.024 mol/l

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR