Palladium(II)klorid (59% Pd) hydrogenation catalyst

Leverandør: VWR Chemicals
Warning

Synonymer: Palladium dichloride

26136.083EA 2375 NOK
26136.083
Palladium(II)klorid (59% Pd) hydrogenation catalyst
Palladium(II)klorid
Formel: PdCl₂
MW: 177.33 g/mol
Smeltepunkt: 678
Tetthet: 4 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003558
CAS-nummer: 7647-10-1
EINECS: 231-596-2
FN: 3260
ADR: 8 III
Merck Indeks: 13,07058

Bestill nå

Spesifikasjon Testresultater

Assay (on dried substance) Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR