Bly(II)nitrat ≥98.5%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

Synonymer: Lead dinitrate

26554.260EA 860 NOK
26554.260
Bly(II)nitrat ≥98.5%, GPR RECTAPUR®
Bly(II)nitrat
Formel: Pb(NO₃)₂
MW: 331.21 g/mol
Smeltepunkt: 450…470 °C
Tetthet: 4.53 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011153
CAS-nummer: 10099-74-8
EINECS: 233-245-9
FN: 1469
ADR: 5.1 (6.1) II
Merck Indeks: 13,05432

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98.5 %
Substances not precipitated by H2SO4 Max. 0.5 %
Cl (Chloride) Max. 50 ppm
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR