Kaliumklorid ≥97%, TECHNICAL

Leverandør: VWR Chemicals

Synonymer: Potassiumchloride

26752.366EA 1175 NOK
26752.366
Kaliumklorid ≥97%, TECHNICAL
Kaliumklorid
Formel: KCl
MW: 74.55 g/mol
Kokepunkt: 1420
Smeltepunkt: 773
Tetthet: 1.98 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8
Merck Indeks: 14,07621

Bestill nå

Spesifikasjon Testresultater

Assay Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR