Ammonium cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Warning

Synonymer: Tetraammonium cerium(IV) sulphate, Cerium (IV) ammonium sulphate, Ammonium ceric sulphate, Ammonium cerium sulphate hydrate, Ammonium cerium(IV) sulphate hydrate, Tetraammonium cerium tetrakis(sulphate)

2678.1000EA 2250 NOK
2678.1000
Ammonium cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Ammonium cerium(IV) sulphate
Formel: H₁₆CeN₄O₁₆S₄
MDL nummer: MFCD00148851
CAS-nummer: 7637-03-8
EINECS: 231-567-4
FN: 3264
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR