Kaliumdikromat ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisk reagens

Leverandør: VWR Chemicals
Danger

26784.231EA 1600 NOK
26784.231 26784.297
Kaliumdikromat ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisk reagens
Kaliumdikromat
Formel: K₂Cr₂O₇
Kokepunkt: 500 °C (1013 hPa)
Smeltepunkt: 398 °C
Tetthet: 2,67 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011367
CAS-nummer: 7778-50-9
EINECS: 231-906-6
FN: 3087
ADR: 5.1 (6.1),II
REACH: 01-2119454792-32
Merck Indeks: 12,07790

Bestill nå

Spesifikasjon Testresultater

Assay (calculated on dried substance) Min. 99.8 %
Insolubility in water Max. 0.03 %
Loss on drying (130°C) Max. 0.05 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 200 ppm
Ca (Calcium) Max. 20 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 10 ppm
Na (Sodium) Max. 0.1 %
Pb (Lead) Max. 50 ppm
Statement on the use Confirmed
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR