Kaliumjodat ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Warning

26839.264EA 3075 NOK
26839.264
Kaliumjodat ≥99%, GPR RECTAPUR®
Kaliumjodat
Formel: KlO₃
MW: 214 g/mol
Smeltepunkt: 560
Tetthet: 3.98 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011406
CAS-nummer: 7758-05-6
EINECS: 231-831-9
FN: 1479
ADR: 5.1 II
Merck Indeks: 13,07726

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Br + Cl + ClO3 (as Cl) Max. 0.05 %
SO4 (Sulphate) Max. 0.02 %
Fe (Iron) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR