≥96%, TECHNICAL

Leverandør: VWR Chemicals

Synonymer: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

26927.460EA 6850 NOK
26927.460
≥96%, TECHNICAL
Kaliumhydrogenortofosfat
Formel: K₂HPO₄
MW: 174.18 g/mol
Smeltepunkt: 340
Tetthet: 2.3 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011383
CAS-nummer: 7758-11-4
EINECS: 231-834-5
Merck Indeks: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR