Sodium-3-(trimethylsilyl)propionate-[D₄], Supelco®

Leverandør: Merck

Synonymer: 2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, Sodium 3-(trimethylsilyl)propionate-[²H₄], 3-(Trimethylsilyl)propionic acid-d4 sodium salt

M-Clarity™ quality level = MQ100

269913-1G
269913-1GEA 3325 NOK
269913-1G
Sodium-3-(trimethylsilyl)propionate-[D₄], Supelco®
Sodium-3-(trimethylsilyl)propionate-[D₄]
Formel: C₆H₉D₄NaO₂Si
MW: 172.21 g/mol
MDL nummer: MFCD00002762
CAS-nummer: 24493-21-8
EINECS: 246-286-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27dd9946734c55d030c348505e9a75af5a733c7&vid=56c03cdf6a5f368e7aa5540ad91573c046c23c01&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR