VWR®, Protein marker III (pre-stained), peqGOLD
Leverandør: VWR Chemicals
27-1110EA 900 NOK
27-1110
VWR®, Protein marker III (pre-stained), peqGOLD
Elektroforese reagenser Molekylvekt markører
Protein Marker III for monitoring protein separation, transfer efficiency and size determination.
  • Mix of six highly purified proteins
  • Range: 20 to 120 kDa
  • Concentration: 200 ng/µl each protein
  • Each protein covalently pre-stained with a blue colouring
  • Ready to use for a fast, convenient workflow