Pyridin, Wasser_0,01 99,5, AnalaR NORMAPUR® zA

Leverandør: VWR Chemicals
Danger

Synonymer: Azabenzene, Azine

27197.295EA 2750 NOK
27197.295
Pyridin, Wasser_0,01 99,5, AnalaR NORMAPUR® zA
Pyridine
Formel: C₅H₅N
MW: 79.1 g/mol
Kokepunkt: 115
Smeltepunkt: –44 °C
Tetthet: 0.983 g/cm³ (20 °C)
Flash punkt: 17
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011732
CAS-nummer: 110-86-1
EINECS: 203-809-9
FN: 1282
ADR: 3 II
Merck Indeks: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colourless liquid
Assay (calculated on anhydrous) Min. 99.5 %
Acidity or alkalinity Max. 0.002 meq/g
Water (K.F.) Max. 0.0100 %
Residue on evaporation Max. 0.0010 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR