Resorcinol 99.0-100.5%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Warning

Synonymer: 1,3-Dihydroxybenzene, 1,3-Benzenediol

27378.234EA 1575 NOK
27378.234
Resorcinol 99.0-100.5%, AnalaR NORMAPUR® analytical reagent
Resorcinol
Formel: C₆H₆O₂
MW: 110.11 g/mol
Kokepunkt: 281
Smeltepunkt: 109…111 °C
Tetthet: 1.272 g/cm³ (20 °C)
Flash punkt: 127 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002269
CAS-nummer: 108-46-3
EINECS: 203-585-2
FN: 2876
ADR: 6.1 III
Merck Indeks: 12,08323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fa2900537bdb42ccfd55e4318288eb881a61a0&vid=e62c617319ff4711a7ff00e69774ad92cfd75d09&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 100.5 %
Acidity or alkalinity Max. 0.005 meq/g
Melting point 109 - 112 °C
Ignition residue (SO4) Max. 0.02 %
Insolubility in water Max. 50 ppm
Insolubility in methanol Max. 50 ppm
Phenol 0.02%
Pyrocatechol 0.02%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR