(S)-(-)-β-Citronellol, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

27483-1ML-F 27483-5ML-F
27483-1ML-FEA 1125 NOK
27483-1ML-F 27483-5ML-F
(S)-(-)-β-Citronellol, Supelco®
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Tetthet: 0,855 g/cm³ (20 °C)
MDL nummer: MFCD00063214
CAS-nummer: 7540-51-4
EINECS: 231-415-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR