Silikagel, hvit, small, pored beads 2,5 - 4 mm

Leverandør: VWR Chemicals

27612.292EA 715 NOK
27612.292
Silikagel, hvit, small, pored beads 2,5 - 4 mm
Silikagel

Glassy, hard beads with an internal surface of approx. 800 m²/g. Because of its very large surface area, our beads exhibit a high adsorption capacity for water vapour. The beads can be reactivated without significantly reducing adsorption efficiency.


  • Very economical
  • Easy to dispose of
  • Without any known adverse effects on the environment

Formel: SiO₂
MW: 60,08 g/mol
Kokepunkt: 2230 °C (1013 hPa) (lit.)
Smeltepunkt: > 1600 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 7631-86-9

Bestill nå

Spesifikasjon Testresultater

Bulk density 680 - 780 g/L
Adsorption capacity (H₂O; 23 °C; 80% RH) Min. 21,5%
Moisture (loss on drying (140 °C) Max. 2,0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR