Silikagel, granules, TECHNICAL
Leverandør: VWR Chemicals
27613.294EA 655 NOK
27613.294
Silikagel, granules, TECHNICAL
Silikagel
Moisture absorber, neutral, not deliquescing, not corrosive. It can be regenerated by heating at about 150 °C
Formel: SiO₂
Kokepunkt: 2230 °C (1013 hPa)
Smeltepunkt: > 1600 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 63231-67-4
Bestill nå
Spesifikasjon Testresultater
Loss on drying (140 °C; 4 h) Max. 3%
Water-absorption capacity (40% RH) Min. 20%