Natriumbromat ≥98%, TECHNICAL

Leverandør: VWR Chemicals
Danger

27734.460EA 0 NOK
27734.460
Natriumbromat ≥98%, TECHNICAL
Sodium bromate
Formel: BrNaO₃
Smeltepunkt: 381 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003476
CAS-nummer: 7789-38-0
EINECS: 232-160-4
FN: 1494
ADR: 5.1,II
Merck Indeks: 12,08736

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR