Natriumbromid ≥98%, GPR RECTAPUR®

Leverandør: VWR Chemicals

27742.290EA 890 NOK
27742.290
Natriumbromid ≥98%, GPR RECTAPUR®
Sodium bromide
Formel: NaBr
MW: 102.89 g/mol
Kokepunkt: 1396
Smeltepunkt: 753
Tetthet: 3.2 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003475
CAS-nummer: 7647-15-6
EINECS: 231-599-9
REACH: 01-2119490106-41
Merck Indeks: 13,08666

Bestill nå

Spesifikasjon Testresultater

Assay (calculated on dried substance) Min. 98 %
Heavy metals (as Pb) Max. 50 ppm
Loss on drying (150°C) Max. 2 %
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR