Natriumklorid ≥98%, TECHNICAL

Leverandør: VWR Chemicals

27788.297EA 230 NOK
27788.297 27788.366 27788.515 27788.460
Natriumklorid ≥98%, TECHNICAL
Natriumklorid
Formel: NaCl
Kokepunkt: 1413 °C (1013 hPa)
Smeltepunkt: 801 °C
Tetthet: 2,16 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3
Merck Indeks: 14,08599

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98 %
Appearance Colourless crystals


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR