tri-Natriumcitrat dihydrat ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals

27830.294EA 680 NOK
27830.294 27830.363 27830.460
tri-Natriumcitrat dihydrat ≥99%, GPR RECTAPUR®
tri-Natriumcitrat dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Smeltepunkt: 150 °C
Tetthet: 1,76 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150031
CAS-nummer: 6132-04-3
EINECS: 200-675-3
REACH: 01-2119457027-40
Merck Indeks: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride) Max. 50 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR