Natriumtetrahydroborat ≥97%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

27881.130EA 525 NOK
27881.130 27881.180
Natriumtetrahydroborat ≥97%, GPR RECTAPUR®
Sodium borohydride
Formel: NaBH₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003518
CAS-nummer: 16940-66-2
EINECS: 241-004-4
FN: 1426
ADR: 4.3,I

Bestill nå

Spesifikasjon Testresultater

Assay min. 97%
Identity passes test
Appearance white hygroscopic crystals


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR