Natriumtiosulfat pentahydrat ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals

27909.296EA 570 NOK
27909.296 27909.365 27909.460
Natriumtiosulfat pentahydrat ≥99%, GPR RECTAPUR®
Natriumtiosulfat pentahydrat
Formel: Na₂S₂O₃.5H₂O
Smeltepunkt: 48 °C
Tetthet: 1,67 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149186
CAS-nummer: 10102-17-7
EINECS: 231-867-5
Merck Indeks: 12,08844

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 100 ppm
Fe (Iron) Max. 10 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR