Natriummetafosfat, GPR RECTAPUR®

Leverandør: VWR Chemicals

27930.290EA 560 NOK
27930.290
Natriummetafosfat, GPR RECTAPUR®
Natriummetafosfat
Formel: NaPO₃
MW: 101.96 g/mol
Smeltepunkt: 625
Tetthet: 2.54 g/cm³ (20 °C)
CAS-nummer: 10361-03-2
EINECS: 233-782-9

Bestill nå

Spesifikasjon Testresultater

di-Phosphorus pentoxide Min. 60 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 0.05 %
Fe (Iron) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR