Oppsamlingsplater
Leverandør: Cytiva
28-4039-43
28-4039-43EA 590 NOK
28-4039-43
Oppsamlingsplater
Mikroplater HTS-mikroplater
Made from biocompatible PP. Collection plates, 96-well, designed for collection of eluted samples in parallel from screening plates such as MultiTrap™ and PreDictor™ plates.
  • Convenient collection of samples in 96-well format
  • Excellent fit to prepacked MultiTrap plates and PreDictor plates
  • V-bottom minimises the loss of sample
Well volume: < 500 µl