Hydrophobic interaction chromatography columns, HiPrep™ Phenyl FF
Leverandør: Cytiva
28-9365-45
28-9365-45EA 8425 NOK
28-9365-45
Hydrophobic interaction chromatography columns, HiPrep™ Phenyl FF
Kromatografikolonner
HiPrep™ Phenyl FF 16/10, are prepacked Phenyl Sepharose™ 6 Fast Flow columns ready to use for hydrophobic interaction chromatography (HIC).
  • 20 ml HIC columns provide fast, preparative separation of proteins and other biomolecules
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations
  • Phenyl Sepharose™ 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity
  • Two levels of substitution help find optimal selectivity and binding capacity