Hydrophobic interaction chromatography column, HiScreen™ Phenyl HP
Leverandør: Cytiva
28-9505-16
28-9505-16EA 2000 NOK
28-9505-16
Hydrophobic interaction chromatography column, HiScreen™ Phenyl HP
Kromatografikolonner
HiScreen Phenyl HP columns are prepacked with Phenyl Sepharose High Performance hydrophobic interaction chromatography (HIC) resins. The columns are an excellent choice for method optimization and parameter screening.
  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to