MagRack™ Maxi
Leverandør: Cytiva
28-9864-41
28-9864-41EA 6325 NOK
28-9864-41
MagRack™ Maxi
Stativer for rør Magnetisk seperasjonsstativ
MagRack Maxi is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads used for sample volumes up to 50 ml.
  • Provides convenience by making it easy to perform small-scale purification and enrichment of sample volumes up to 50 ml
  • Allows for optimal performance during protein purifications or enrichments
  • Each rack consists of an aluminium housing (blue) and a detachable plastic bar (white) containing a neodymium magnet
The anodized aluminium housing assists with electrical conductivity whilst the neodymium magnetMore Product Information