Hydrophobic interaction chromatography columns, HiScreen™ Capto™ Butyl
Leverandør: Cytiva
28-9924-73
28-9924-73EA 1975 NOK
28-9924-73
Hydrophobic interaction chromatography columns, HiScreen™ Capto™ Butyl
Kromatografikolonner
Capto Phenyl (High sub) and Capto Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
  • Prepacked with Capto Butyl
  • Excellent for method optimization and parameter screening due to the 10 cm bed height
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same resin using the same flow velocity
HiScreen columns are part of the processMore Product Information