tetra-Natriumdifosfat dekahydrat ≥97%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Warning

28053.295EA 1075 NOK
28053.295
tetra-Natriumdifosfat dekahydrat ≥97%, GPR RECTAPUR®
tetra-Natriumdifosfat dekahydrat
Formel: Na₄P₂O₇·10H₂O
Smeltepunkt: 990 °C
Tetthet: 1,82 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149200
CAS-nummer: 13472-36-1
EINECS: 231-767-1

Bestill nå

Spesifikasjon Testresultater

Assay Min. 97 %
Heavy metals (as Pb) Max. 20 ppm
SO4 (Sulphate) Max. 0.05 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR