Natriumsulfat ≥99.5%, PESTINORM® for pesticide residue analysis
Leverandør: VWR Chemicals
Synonymer: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
28116.293EA 1500 NOK
28116.293
Natriumsulfat ≥99.5%, PESTINORM® for pesticide residue analysis
Sodium sulphate
Formel: Na₂SO₄
Kokepunkt: 1429 °C (1013 hPa)
Smeltepunkt: 888 °C
Tetthet: 2,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003504
CAS-nummer: 7757-82-6
EINECS: 231-820-9
Merck Indeks: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Assay Min. 99.5 %
Absence for pesticide residues Passes test
pH (20°C; 5 %) 5.0 - 8.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Loss on drying (130°C) Max. 0.5 %
Loss on ignition (600°C) Max. 0.5 %
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 10 ppm
PO4 (Phosphate) Max. 20 ppm
As (Arsenic) Max. 1 ppm
Ca + Mg (as Ca) Max. 100 ppm
Fe (Iron) Max. 2 ppm
K (Potassium) Max. 100 ppm