Strontium klorid heksahydrat ≥99.0%, AnalaR NORMAPUR® analytical reagent
Leverandør: VWR Chemicals
Warning
28321.268EA 1550 NOK
28321.268
Strontium klorid heksahydrat ≥99.0%, AnalaR NORMAPUR® analytical reagent
Strontium klorid heksahydrat
Formel: SrCl₂·6H₂O
MW: 266.62 g/mol
Kokepunkt: 100
Smeltepunkt: 61
Tetthet: 1.93 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149865
CAS-nummer: 10025-70-4
EINECS: 233-971-6
Merck Indeks: 13,08920
Bestill nå
Spesifikasjon Testresultater
Assay Min. 99.0 %
pH (20°C; 5 %) 5.0 - 7.0
Insolubility in water Max. 50 ppm
NO3 (Nitrate) Max. 20 ppm
SO4 (Sulphate) Max. 25 ppm
Ba (Barium) Max. 50 ppm
Ca (Calcium) Max. 0.05 %
Cu (Copper) Max. 2 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 100 ppm
Na (Sodium) Max. 150 ppm
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 2 ppm