Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol 90% (v/v)/methanol 10% (v/v), AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Danger

28533.261EA 1025 NOK
28533.261
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol 90% (v/v)/methanol 10% (v/v), AVS TITRINORM volumetric solution
Tetrabutylammonium hydroxide
Formel: C₁₆H₃₇NO
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009425
CAS-nummer: 2052-49-5
EINECS: 218-147-6
FN: 1992
ADR: 3 (6.1),II

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR