Dietyleter ≥99.5% (by GC, corrected for water content) stabilised, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

Synonymer: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilisering: 1.5 - 2.5% Stabilised with ethanol

2854-25EA 995 NOK
2854-25
Dietyleter ≥99.5% (by GC, corrected for water content) stabilised, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Dietyleter

Especially intended for use as an HPLC and TLC solvent. Stabilized with ethanol. Filtered through a 0.2 µm element.

Formel: (CH₃CH₂)₂O
MW: 74.12 g/mol
Kokepunkt: 35 °C (1013 hPa)
Smeltepunkt: –123 °C
Tetthet: 0.7135 g/cm³ (20 °C)
Flash punkt: –40 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011646
CAS-nummer: 60-29-7
EINECS: 200-467-2
FN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Indeks: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 99.5 %
Assay ((C₂H₅)₂O) (by GC, exclusive of preservative, corrected for water) ≥99.5 %
Alcohol (C₂H₅OH)(by GC) (preservative)(v/v) 1.5 - 2.5 %
Color (APHA) ≤10
Peroxide (as H₂O₂)(at time of packaging) ≤0.0005 %
Optical Absorbance (1-cm path vs water) - 215 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 230 nm ≤0.30
Optical Absorbance (1-cm path vs water) - 254 nm ≤0.08
Optical Absorbance (1-cm path vs water) - 280 nm ≤0.01
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.005
Residue after Evaporation ≤0.0005 %
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR