4,4'-Bi-o-toluidin 0,1% 1.6 M hydrochloric solution for chlorine determination in water according to the NF T 90-010 standard

Leverandør: VWR Chemicals
Danger

Synonymer: 4,4'-Bi-o-toluidine

28672.294EA 1600 NOK
28672.294
4,4'-Bi-o-toluidin 0,1% 1.6 M hydrochloric solution for chlorine determination in water according to the NF T 90-010 standard
4,4'-Bi-o-toluidin

Determination of oxygen or free chlorine

Formel: [-C₆H₃(CH₃)-₄-NH₂]₂
MDL nummer: MFCD00014773
CAS-nummer: 119-93-7
EINECS: 204-358-0
FN: 1760
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR