2',7'-Dichlorofluorescin diacetate ≥94% (by HPLC), Calbiochem®

Leverandør: Merck Millipore Calbiochem


Packaged under inert gas.

287810-100
287810-100EA 2375 NOK
287810-100
2',7'-Dichlorofluorescin diacetate ≥94% (by HPLC), Calbiochem®
2',7'-Dichlorofluorescin diacetate

A cell permeable fluorogenic probe used for the detection of reactive oxygen species (ROS) and nitric oxide (NO) and for the determination of the degree of overall oxidative stress.

Formel: C₂₄H₁₆Cl₂O₇
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00128955
CAS-nummer: 4091-99-0

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) ≥94%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR