Polysorbat 80, GPR RECTAPUR®

Leverandør: VWR Chemicals

28830.291EA 1075 NOK
28830.291 28830.260 28830.320 28830.460
Polysorbat 80, GPR RECTAPUR®
Polysorbat 80
Kokepunkt: >100 °C (1013 hPa)
Smeltepunkt: –21 °C
Tetthet: 1,062 g/cm³ (20 °C)
Flash punkt: 149 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082107
CAS-nummer: 9005-65-6
EINECS: 500-019-9
Merck Indeks: 13,07664

Bestill nå

Spesifikasjon Testresultater

Identification Passes test
Density (20/4) 1.070 - 1.090
Hydroxyl value 65 - 80
Saponification value 45 - 55
Acid value Max 2 mg KOH/g
Water content Max 3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR