Silikagel pore size 60 Å, 2-22 μm particle size distribution, without binder, high purity suitable for thin layer chromatography (TLC), Supelco®

Leverandør: Merck

288500-1KG
288500-1KGEA 1725 NOK
288500-1KG
Silikagel pore size 60 Å, 2-22 μm particle size distribution, without binder, high purity suitable for thin layer chromatography (TLC), Supelco®
Silikagel
Formel: SiO₂
Kokepunkt: 2230 °C
Smeltepunkt: > 1600 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 63231-67-4
EINECS: 231-545-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR