Vanadylsulfat pentahydrat, TECHNICAL

Leverandør: VWR Chemicals
Warning

Synonymer: Basic vanadium (IV) sulphate pentahydrate, Vanadium oxide sulphate pentahydrate, Vanadium (IV) sulphate oxide pentahydrate

28900.180EA 9900 NOK
28900.180
Vanadylsulfat pentahydrat, TECHNICAL
Vanadylsulfat pentahydrat
Formel: VOSO₄·5H₂O
Tetthet: 2 g/cm³ (20 °C)
MDL nummer: MFCD00149933
CAS-nummer: 12439-96-2
EINECS: 248-652-7
FN: 2931
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Identification Passes test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR