Untagged chromatography column package
Leverandør: Cytiva
29-4976-31
29-4976-31EA 10250 NOK
29-4976-31
Untagged chromatography column package
Kromatografikolonner
The Untagged column package includes cation/anion exchange and size exclusion chromatography columns for purification of untagged/native proteins. The columns provided in the package are prepacked with Capto Q ImpRes and Capto SP ImpRes high-performance resins and can be used individually or in combination.
  • Resins provide you with excellent pressure/flow properties and small resin bead sizes to maximize resolution
  • The resins give you high loading capacities over a broad pH range
Final high purity is obtained with HiPrep 16/60 Sephacryl S-200 HR, aMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Oppbevaring 4...30 °C