TEV Protease Kit
Leverandør: Biotium
29090-1000U
29090-1000UEA 875 NOK
29090-1000U
TEV Protease Kit
Proteinrense Protein Purification Kits
TEV protease kit that may be used for cleaving proteins or peptides containing the TEV protease recognition sequence E-N-L-Y-F-Q↓(S/G/A/M/C/H).
TEV protease is a highly sequence-specific cysteine protease from Tobacco Etch Virus (TEV). The TEV Protease recognition sequence with the highest catalytic efficiency is the sequence Glu-Asn-Leu-Tyr-Phe-Gln-Ser with optimal cleavage occurring between the Gln and Ser residues. However, the amino acid in the P1’ position (amino acid immediately downstream of the cleavage site) can also be Gly, Ala, Met, Cys, or His. The protease is used to cleave affinity tags such as malt ose-binding protein (MBP) or poly-histidine from fusion proteins. The optional temperature for cleavage is 30 °C but it can also be used at temperatures as low as 4 °C.
Recombinant TEV is expressed from E. coli as a single, non-glycosylated polypeptide chain with N-terminal 7X His-tag for easy removal from the reaction using nickel affinity chromatography. The TEV kit control Substrate as an optional reaction control. It is a His-tagged fusion protein (MBP-TEV substrate-His6) around 80,3 kDa, observed by reducing SDS-PAGE. When Control Substrate is cleaved with TEV, two bands at 43,3 kDa and 37 kDa are observed under reducing SDS-PAGE.
Control substrate included for assessing reaction efficiency by SDS-PAGE.
Leveringsinformasjon: The kit includes recombinant TEV expressed from E. coli, TEV Protease with N-terminal 7X His-tag, Control substrate (MBP-TEV substrate-His6), and 10X Reaction Buffer.