Zinkklorid 71% ≥71%

Leverandør: VWR Chemicals
Danger

Synonymer: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

29142.290EA 3300 NOK
29142.290
Zinkklorid 71% ≥71%
Zinkklorid
Formel: ZnCl₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011295
CAS-nummer: 7646-85-7
EINECS: 231-592-0
FN: 1840
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Assay Min. 71 %
Heavy metals (as Pb) Max. 20 ppm
SO4 (Sulphate) Max. 0.02 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR