Reclosable fastener, SJ457D, Dual Lock™
Leverandør: 3M
SJ457D
293-0013EA 2375 NOK
293-0013
Reclosable fastener, SJ457D, Dual Lock™
Tape
3M™ Dual Lock™ low profile reclosable fastener SJ4570 uses a modified acrylic adhesive on the back of a clear polypropylene reclosable fastener. This modified acrylic adhesive enables bonding of medium to low surface energy materials such as metals, glass, many plastics, and powder coated paints.
  • Half the thickness (1,7 mm) of standard Dual Lock™ Reclosable Fasteners for low profile attachments
  • Opens and closes up to 100 times before losing 50% of original tensile strength
  • Translucent fastener and adhesive is ideal for joining transparent materials